PharmaMOP™ SingleSTEP Sahara™ Mop Head
PharmaMOP™ SingleSTEP Sahara™ Mop Head
Actual product may vary
PharmaMOP™ SingleSTEP Sahara™ Mop Head
Designed for weekly cleaning, this mop strips heavy soils and residues from floors, walls, stainless steel, and windows while allowing even application of disinfectants
Have Questions?
Catalog number FOAFS859SH
Price (MYR)
5,129.00
CS
Add to cart
Price (MYR)
5,129.00
CS
Add to cart
  • Designed for walls, floors, and ceilings in ISO Class 1-7 clean rooms
  • Ultra lightweight
  • Two axis swivel reduces work load
  • Individually packed
Specifications
ColorWhite, Green (Foam Strips)
Size0.25 x 5 x 16 in.
For Use With (Application)Designed for walls, floors, and ceilings in ISO Class 1-7 clean rooms
MaterialPolyester Microfiber
Unit SizeCS