Check Order Status
Hydro H 11 V Vaporization Water Bath with BS1363 UK-Plug
Actual product may vary
Hydro H 11 V Vaporization Water Bath with BS1363 UK-Plug
Vaporization water bath for gentle steaming work
Have Questions?
Catalog number
FKS16679385
Price (MYR)
14,602.00
EA
Add to cart
Price (MYR)
14,602.00
EA
Add to cart
Temperature adjustment via rotary knob with temperature scale
1-point temperature controller
Water deficiency protection
Adjustable water level regulator, sieve bottom and ring set as standard
Bath interior, cover, sieve plate and heating element made of stainless steel
All hole covers consist of a multi-part ring set and are removable
Multi-piece ring set made of heat-resistant plastic
Internal hole provided on cover frame to accommodate a stand rod
Technical Features:
Working temperature range: 25 to 100 °C
Ambient temperature range: 10 to 40 °C
Temperature stability: 3 ± K
Heater power max: 1.5 kW
Power consumption max: 1.5 kW
Power consumption: 13 A
Size of bath (Ø): 91 mm
Number of bath openings: 6
Overall dimensions (W x D x H): 450 x 300 x 155 mm
Power supply: 230 V, 50/60 Hz
Power plug: power cord with angled plug (BS1363)
Specifications
Temperature Range
25°C to 100°C
Voltage
230 V