Check Order Status
Whatman™ Nuclepore Hydrophilic Membrane
Actual product may vary
Whatman™ Nuclepore Hydrophilic Membrane
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
Have Questions?
Catalog Number
Pore Size
Diameter (Metric)
FKS16645665
3 μm
13 mm
Catalog number
FKS16645665
Price (MYR)
825.00
PK
Add to cart
Pore Size:
3 μm
Diameter (Metric):
13 mm
Price (MYR)
825.00
PK
Add to cart
Track-etched for highly defined pore size and high pore density
Treated with PVP to make it hydrophilic
Low extractables minimizing filtration sample contamination risk
High chemical resistance
Good thermal stability
Low, consistent ash
Low tare weights
Smooth, flat membrane surface for high particle visibility
Applications:
Bioassays
Cell biology
Environmental analysis
Microscopy
Cytology
Parasitology
Certain air and oceanography analyses
Specifications
Material
Polycarbonate
Pore Size
3 μm
For Use With (Application)
Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses
Quantity
100 Pk.
Type
Nuclepore hydrophilic membrane
Diameter (Metric)
13 mm