Reusable Glass Heavy-Duty Low-Form Beakers
Reusable Glass Heavy-Duty Low-Form Beakers
Actual product may vary
Reusable Glass Heavy-Duty Low-Form Beakers
Reusable Glass Heavy-Duty Low-Form Beakers
Reusable Glass Heavy-Duty Low-Form Beakers
Spout designed for optimum performance
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS165858301000 mL
FISFB-101-600600 mL
FISFB10110001000 mL
Catalog number FKS16585830
Price (MYR)
1,465.00
PK
Add to cart
Capacity (Metric):
1000 mL
Price (MYR)
1,465.00
PK
Add to cart
Fisherbrand™ Reusable Glass Heavy-Duty Low-Form Beakers feature a sturdy construction ideal for mechanical shock and other harsh applications. Spout is designed for optimum performance.
  • Thick uniform walls
  • Evenly tooled top rim with extra wall thickness
  • Durable matte finish marking area
  • White graduated scale
Specifications
Accuracy±5%
Capacity (English)33.81 oz.
Diameter (English) Outer4.21 in.
Diameter (Metric) Outer10.7 cm
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
For Use With (Application)Offer Superior Mechanical Strength and Durability, Provides Improved Safety When Used Under Extreme Conditions Such as Mechanized Washing and Rough Handling
Graduation Range100 to 1000 mL
Height (English)5.98 in.
Height (Metric)15.2 cm
IncludesSpout
Increments50 mL
Label TypeMatte Finish Marking
Quantity6 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960, ASTM E438, USP Type I
ScaleDual Scale
Unit SizePK