Borosilicate Glass Colorimetry Cells
Borosilicate Glass Colorimetry Cells
Actual product may vary
Borosilicate Glass Colorimetry Cells
Precision fused cells
Have Questions?
Catalog NumberFor Use With (Equipment)Pathlength
FKS16536640Model Fs/PFX195/880/950/99510 mm
Catalog number FKS16536640
Price (MYR)
-
For Use With (Equipment):
Model Fs/PFX195/880/950/995
Pathlength:
10 mm
Available in a range of dimensions and path lengths, made from optical glass to the highest standards. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.
Series: W600 Borosilicate
Specifications
TypeColorimetry Cell
For Use With (Equipment)Model Fs/PFX195/880/950/995
MaterialBorosilicate Glass
Pathlength10 mm
Unit SizeEA