White Disposable Forceps
White Disposable Forceps
Actual product may vary
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
White Disposable Forceps
Disposable forceps are ideal for grasping, lifting, and moving small objects like fine tissue or solid samples.
Have Questions?
Catalog NumberTip StyleSterility
FKS16535210Angled, PointedSterile
Catalog number FKS16535210
Price (MYR)
917.00
PK
Add to cart
Tip Style:
Angled, Pointed
Sterility:
Sterile
Price (MYR)
917.00
PK
Add to cart
  • The fluted handles ensure optimum and secure handling as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Sterile forceps are individually wrapped
  • For single use
  • Cleanroom manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Tip StyleAngled, Pointed
No. per Pack100 (packaged individually)
Length (English)5.12 in.
MaterialPolystyrene
Length (Metric)130 mm
Product TypeForceps
SterilitySterile
StyleStandard
Unit SizePK