Wire Drying Rack
Wire Drying Rack
Actual product may vary
Wire Drying Rack
Wire Drying Rack
Wire Drying Rack
Wire Drying Rack
The Wire Drying Rack saves glassware from chips and cracks. Test tubes, beakers, flasks, and other labware can be drained and dried on the rack.
Have Questions?
Catalog NumberPositions
FKS1633074690
Catalog number FKS16330746
Price (MYR)
1,839.00
EA
Add to cart
Positions:
90
Price (MYR)
1,839.00
EA
Add to cart
  • Self-standing rack
  • Place on a table or near a sink
  • Use hooks to hang rack on the wall
  • Holds up to 90 pieces of labware
  • Pin Sizes: 18 x 127 mm, and 72 x 76 mm
  • Constructed of spot-welded, vinyl-coated steel wire for durability
  • Polystyrene tray and back
  • Easily accessible and removable bottom drainage pan
Specifications
IncludesDrip Pan
Product TypeDrying Rack
DescriptionWire Drying Rack
AutoclavableNot Autoclavable
ColorBlack/Gray
HoldsLab Glassware
MaterialVinyl-coated Steel Wire
Positions90
StackableNo
SterilityNon-sterile
ModelHS23243A
No. per Case2/Cs.
No. per Pack1 Ea.
Dimensions (L x W x H)18.2 x 7.4 x 20.7 in.
Length (Metric)46.2 cm
Length (English)18.2 in.
Width (Metric)18.2 cm
Width (English)7.4 in.
Height (Metric)52.5 cm
Height (English)20.7 in.
Weight (English)5.62 lb.
Weight (Metric)2.53 kg
Unit SizeEA