Check Order Status
Plain PTFE Stir Bars
Actual product may vary
Plain PTFE Stir Bars
Provides stable stirring action. Fisherbrand™ Plain PTFE Stir Bars provide greater turbulence than cylindrical stir bars.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
Quantity
FKS16295791
50 mm
8 mm
2/Pk.
FKS16235791
40 mm
8 mm
5/Pk.
FKS16295781
30 mm
6 mm
5/Pk.
3 Options
Catalog number
FKS16295791
Price (MYR)
293.00
PK
Add to cart
Length (Metric):
50 mm
Diameter (Metric):
8 mm
Quantity:
2/Pk.
Price (MYR)
293.00
PK
Add to cart
Stable, general-purpose magnetic stir bar
Provides greater turbulence than cylindrical stir bars
Autoclavable
Specifications
Autoclavable
Autoclavable
Certifications/Compliance
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color
White
Description
PTFE encapsulated Alnico V magnet core
For Use With (Equipment)
Containers & Vessels / Stirrer Hot Plates
Coating
PTFE
Diameter (Metric)
8 mm
Length (Metric)
50 mm
Quantity
2/Pk.
Type
PTFE Stirrer Bars Plain