Volac™ Glass Pasteur Pipets
Volac™ Glass Pasteur Pipets
Actual product may vary
Volac™ Glass Pasteur Pipets
Volac™ Glass Pasteur Pipets
Volac™ Glass Pasteur Pipets
Volac™ Glass Pasteur Pipets
Use these pipets for a variety of applications. Poulten Graf™ Volac™ Glass Pasteur Pipets feature a slightly heavier wall thickness to provide greater strength and reduce the risk of breakage in use or when fitting PVC teats.
Have Questions?
Catalog NumberLength (Metric)SterilityIncludes
FKS15962290150 mmSterilePlugged
Catalog number FKS15962290
Price (MYR)
-
Length (Metric):
150 mm
Sterility:
Sterile
Includes:
Plugged
  • Available with or without plug
  • Glass pipets come in 150, 230 and 270mm lengths
  • Packing materials are carefully chosen to ensure ease of use for the technician with maximum protection from breakage during transit and minimum lint or dust contamination
  • All standard pipettes can be supplied with 'sealed points' and break mark
Specifications
Certifications/ComplianceBS5732,ISO7712
MaterialSoda Lime Glass
SterilitySterile
Drops/mL30 to 35 mL
IncludesPlugged
Quantity250 Pack
Length (Metric)150 mm
TypePasteur Pipet