Electronic Balance
Electronic Balance
Actual product may vary
Fisherbrand
Electronic Balance
Electronic Balance designed to handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Have Questions?
Catalog NumberCapacityModel
FKS15959345220 gFPOS222
Catalog number FKS15959345
Price (MYR)
3,225.00
EA
Add to cart
Capacity:
220 g
Model:
FPOS222
Price (MYR)
3,225.00
EA
Add to cart
  • Essential weighing applications at a competitive price point
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance
Specifications
Connection TypeRS232, USB, or Ethernet (available as accessories)
Capacity220 g
Display TypeBacklit LCD
Readability0.010000 g
For Use With (Application)Weighing, Parts Counting, Percent Weighing, Checkweighing
IncludesAC Adapter
Linearity0.020000 g
ModelFPOS222
Repeatability0.010000 g
Stabilization Time1.000 sec.
Temperature Range (English)50°F to 104°F
Temperature Range (Metric)10°C to 40°C
Warranty2 Years
Weighing ModesCarat, Grain, Gram, Newton, Ounce, Ounce Troy, Pennyweight
Dimensions (D x W x H)230 x 204 x 70 mm (9.1 x 8 x 2.8 in.)
Weight (English)2.2 lb.
Weight (Metric)1 kg
Unit SizeEA