PowderThief Sampler 32mm Diameter
PowderThief Sampler 32mm Diameter
Actual product may vary
PowderThief Sampler 32mm Diameter
PowderThief Sampler 32mm Diameter
PowderThief Sampler 32mm Diameter
PowderThief Sampler 32mm Diameter
PowderThief Sampler 32mm Diameter
Extensively used throughout the pharmaceutical, chemical and food industries around the world. Sampling Systems™ PowderThief Sampler 32mm Diameter is suitable for sampling free flowing powders and granules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS158692012000 mm
FKS158492011000 mm
Catalog number FKS15869201
Price (MYR)
10,443.00
EA
Add to cart
Length (Metric):
2000 mm
Price (MYR)
10,443.00
EA
Add to cart
  • Made from high quality 316L stainless steel
  • Wide range of sample sizes
  • Wide range of sampler lengths
  • Easy to use
  • A range of tips are available so that a single sampler can be used to accurately sample a range of different volumes
Specifications
Product TypePowder Sampler
MaterialStainless Steel (316)
Handle MaterialStainless Steel (316)
Diameter (Metric)32 mm
Length (Metric)2000 mm
Unit SizeEA