Cleaning Coupons for Metal
Cleaning Coupons for Metal
Actual product may vary
Fisherbrand
Cleaning Coupons for Metal
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
Have Questions?
Catalog NumberMaterialDimensions (L x W)Finish
FKS15819561Stainless Steel (316)100 x 100 mmBright Polish
Catalog number FKS15819561
Price (MYR)
2,907.00
EA
Add to cart
Material:
Stainless Steel (316)
Dimensions (L x W):
100 x 100 mm
Finish:
Bright Polish
Price (MYR)
2,907.00
EA
Add to cart
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Product TypeCleaning Coupon
Dimensions (L x W)100 x 100 mm
FinishBright Polish
For Use With (Application)Testing and validating cleaning regime
MaterialStainless Steel (316)
Unit SizeEA