Polymethylpentene Forceps
Polymethylpentene Forceps
Actual product may vary
Fisherbrand
Polymethylpentene Forceps
Manufactured with PMP for very good resilience. VITLab™ Polymethylpentene Forceps are ideal for use in bottling and decanting.
Have Questions?
Catalog NumberLength (Metric)
FKS15751996145 mm
Catalog number FKS15751996
Price (MYR)
238.00
PK
Add to cart
Length (Metric):
145 mm
Price (MYR)
238.00
PK
Add to cart
  • White, Pointed, Elastic 
Specifications
Tip StylePointed
MaterialPMP
Length (Metric)145 mm
Product TypeForceps
Quantity (No. per Case)10/Pack
For Use With (Application)For autoclaving and microwaves, bottling and decanting
TypeForceps
Unit SizePK