Check Order Status
Mulitpurpose Sampling Container
Actual product may vary
Mulitpurpose Sampling Container
500 mL sterile multipurpose sampling container.
Have Questions?
Catalog Number
Capacity (Metric)
FKS15743657
500 mL
Catalog number
FKS15743657
Price (MYR)
1,569.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
1,569.00
PK
Add to cart
Batch and expiry details are stated on the side of the label
Recommended for physical or chemical testing
Ideal for water sampling
Shelf Life: 2 years
Sterilisation Method: Gamma Irradiation to achieve a SAL of 10
-6
Specifications
Material
HDPE
Capacity (Metric)
500 mL
Color
Clear Bottle, Blue Cap
For Use With (Application)
Physical or chemical testing; water sampling
Quantity
40