Expert pH Pocket Tester
Expert pH Pocket Tester
Actual product may vary
Expert pH Pocket Tester
Expert pH Pocket Tester
Expert pH Pocket Tester
Expert pH Pocket Tester
Fisherbrand
Expert pH Pocket Tester
Perform on-the-go, fuss-free measurements for applications such as water treatment, environmental monitoring, education, hydroponics, agriculture, aquaculture, food & beverage and many more!
Have Questions?
Catalog NumberMeasurement Modes
FKS15692047pH, Temperature
Catalog number FKS15692047
Price (MYR)
967.00
EA
Add to cart
Measurement Modes:
pH, Temperature
Price (MYR)
967.00
EA
Add to cart
The Thermo Scientific™ Expert pH Pocket Tester is designed for on-the-go measurements of pH and temperature. The Expert pH Pocket Tester is best used in applications such as hydroponic gardening, aquaculture, agriculture, pools, cooling towers, and more!
  • Large 4 digit double line display
  • Simultaneous display of pH and temperature
  • Battery life, ready, and calibration indicators
  • Automatic temperature compensation
  • Auto buffer recognition and calibration
  • Multi language operational instructions
Display:
  • Large 4 digit double line display and wide viewing angle make it easy to read and record measurements
  • Battery life, ready, and calibration indicators
  • Simultaneous display of pH with temperature
Keypad:
  • Easy to operate and intuitive user interface with power, calibration, menu, and enter keys
  • One push calibration: immerse sensor in calibration solution and press Cal
  • Auto pH buffer recognition and calibration
Cap:
  • Gasketed and leak proof with an O-ring seal
  • Transparent cap with a fill line indicator
  • Symmetrical cap can be front or rear facing
  • Cap secures to the top of tester when not in use
Additional features:
  • Lanyard provision enables the user to easily transport the pocket tester for on-the-go measurements
  • Pocket tester floats and meets Waterproof IP67 rating
  • Wide, stable base allows the tester to independently stand vertically for measurements
  • Automatic temperature compensation: adjusts readings according to temperature change for greater accuracy
  • Auto-off function: pocket tester turns off after 8.5 minutes of non-use to extend battery life
  • CD/RoHS Marking
  • Multi language instructions available
Specifications
Accuracy±0.1 pH
Accuracy (pH)±0.1
Backlight OptionNo
Battery Life100 hr. (average)
Battery Type4 x 1.5V Micro Alkaline
CalibrationpH, temperature
Calibration Points1 to 3
Certifications/ComplianceCE, FCC Class A, ROHS-2
DescriptionExpert pH pocket tester for measuring pH with temperature
Display Type4-digit, Double-line
IP RatingIP67
Measurement ModespH, Temperature
Range (pH)0 to 14
Resolution (pH)0.1
Temperature Range (Metric)0.0°C to 60.0°C
IncludesExpert pH tester with batteries
Relative Humidity Range5 to 85%, non-condensing
Resolution0.1pH
Temperature Accuracy±0.6°C, ±1.0°F
Temperature CompensationYes
Temperature (English) Operating32.0°F to 122.0°F
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point
Temperature Range (English)32.0°F to 140.0°F
Temperature Resolution0.1°C, 0.1°F
Warranty1 Year
Length (English)6.7 in.
Length (Metric)17 cm
Width (English)1.8 in.
Width (Metric)4.5 cm
Height (English)1.2 in.
Height (Metric)3 cm
Weight (English)3.4 oz.
Weight (Metric)96.3 g
Unit SizeEA