Mini Sampler Spoons
Mini Sampler Spoons
Actual product may vary
Mini Sampler Spoons
Avoid cross contamination by using a different spoon for each sample
Have Questions?
Catalog NumberCapacity (Metric)
FKS155901020.5 mL
Catalog number FKS15590102
Price (MYR)
334.00
PK
Add to cart
Capacity (Metric):
0.5 mL
Price (MYR)
334.00
PK
Add to cart
  • These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
  • Made from white high-impact polystyrene plastic
  • All sterile products are lot stamped for accurate tracing
Specifications
Product TypeSpoon
MaterialPolystyrene
AutoclavableNot Autoclavable
IncludesRounded bowl and thin handle neck
Capacity (Metric)0.5 mL
DisposableSingle-use
SterilitySterile
ColorWhite
For Use With (Application)Designed for retrieving small, specific volume samples of powders or similar materials
Unit SizePK