Polygon Magnetic Stirring Bars without Pivot Ring
Polygon Magnetic Stirring Bars without Pivot Ring
Actual product may vary
Fisher Catalogue
Polygon Magnetic Stirring Bars without Pivot Ring
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars
Have Questions?
Catalog number FKS15588682
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification
Specifications
AutoclavableAutoclavable
Certifications/ComplianceISO 9001:2015
ColorWhite
DescriptionStirring Bar
Diameter (English)0.3125 in.
Length (English)0.75 in.
Diameter (Metric)8 mm
Length (Metric)20 mm
MaterialAlnico Magnet
ShapePolygon
Unit SizeEA