Check Order Status
Desiccators with nz 24/29 and Valve
Actual product may vary
Fisherbrand
Desiccators with nz 24/29 and Valve
Made of SIMAX 3.3 borosilicate glass. Fisherbrand Desiccators with nz 24/29 and Valve designed for variety of vacuum work.
Have Questions?
Catalog Number
Product Type
Diameter (Metric) Inner
Diameter (Metric) Outer
Height (Metric)
FKS15544635
Desiccator
192 mm
270 mm
300 mm
Catalog number
FKS15544635
Price (MYR)
3,756.00
EA
Add to cart
Product Type:
Desiccator
Diameter (Metric) Inner:
192 mm
Diameter (Metric) Outer:
270 mm
Height (Metric):
300 mm
Price (MYR)
3,756.00
EA
Add to cart
Made from SIMAX 3.3 borosilicate glass
Designed for variety of vacuum work
Used for desiccation of samples, storage of materials sensitive to humidity
Specifications
Certifications/Compliance
ISO 13 130, DIN 12 491
Material
Simax Borosilicate Glass 3.3
Product Type
Desiccator
Includes
nz 24/29 and valve
Height (Metric)
300 mm
Diameter (Metric) Inner
192 mm
Diameter (Metric) Outer
270 mm