Wrist Motion Shakers
Wrist Motion Shakers
Actual product may vary
Wrist Motion Shakers
Designed to simulate the back and forth motion of the human wrist
Have Questions?
Catalog NumberTimer RangeSpeed RangeDisplayElectrical Requirements
FKS155440700 min. to 99:59 hr.50 to 450 rpmLED display230 V, Plug Type: EU, UK, CN
Catalog number FKS15544070
Price (MYR)
25,024.00
EA
Add to cart
Timer Range:
0 min. to 99:59 hr.
Speed Range:
50 to 450 rpm
Display:
LED display
Electrical Requirements:
230 V, Plug Type: EU, UK, CN
Price (MYR)
25,024.00
EA
Add to cart
  • Adjustable 0-15° mixing angle 
  • Adjustable side arm clamps can accommodate up to 8 flasks (500mL or smaller) 
  • Load capacity of up to 10 lbs. with overload protection 
  • Includes Two 4-position side arm assembly 
  • Cold room compatible ( 5-25°C)
Specifications
Load Bearing Capacity (Metric)4.5 kg
Tilt Angle0 to 15°
Speed Range50 to 450 rpm
Electrical Requirements230 V, Plug Type: EU, UK, CN
Temperature Range (Metric)5°C to 25°C (Cold Room Compatible)
Holds8 x 500 mL/smaller Flasks
Load Bearing Capacity (English)10 lb.
Dimensions (L x W x H)390 x 320 x 300 mm
DescriptionLED display
DisplayLED display
IncludesTwo 4-Position Side Arm Assembly
No. of Positions8
Timer Range0 min. to 99:59 hr.
Unit SizeEA