Check Order Status
316L Stainless Steel Beakers
Actual product may vary
316L Stainless Steel Beakers
Sampling Systems™ 316L Stainless Steel Beakers are ideal for use in the laboratory or production environment.
Have Questions?
Catalog Number
Capacity (Metric)
FKS15529475
1000 mL
Catalog number
FKS15529475
Price (MYR)
1,571.00
EA
Add to cart
Capacity (Metric):
1000 mL
Price (MYR)
1,571.00
EA
Add to cart
Multi purpose container
Ideal for collection and storage of powders and liquids
316L Stainless Steel
Crevice free interior
Available from stock
Polished finish
Specifications
Diameter (Metric) Outer
100mm
Capacity (Metric)
1000 mL
Material
Stainless Steel
Finish
Polished
Height (Metric)
145mm