Check Order Status
Azlon™ USA OSHA Vented Wide Mouth Wash Bottles
Azlon™ USA OSHA Vented Wide Mouth Wash Bottles
Multi-lingual wide neck, OSHA, wash bottles featuring the patented DripLok™ vapor venting valve that prevents solvent drips.
Have Questions?
Catalog Number
Chemical Label
FKS15521232
Acetone
Catalog number
FKS15521232
Price (MYR)
500.00
PK
Add to cart
Chemical Label:
Acetone
Price (MYR)
500.00
PK
Add to cart
Indelibly printed in 3 languages (English, French and Spanish)
Choose from five solvent labels all including the GHS, NFPA diamond, PPE pictograms, risk and safety information
Fitted with color coded one-piece PP closures that maintain their shape in use
Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)
500 mL
Material
Plastic
Quantity
5/Pk.
Chemical Label
Acetone
Closure Type
Vapor-venting