Check Order Status
WIVA Container with Solid Lid
Actual product may vary
WIVA Container with Solid Lid
Remove and dispose of potentially hazardous biological materials safely. Daniels Healthcare™ WIVA Container with Solid Lid conforms to BS 7320 specifications for waste removal.
Have Questions?
Catalog Number
Capacity (Metric)
Dimensions (L x W x H)
FKS15514146
30 L
400 x 335 x 318 mm
Catalog number
FKS15514146
Price (MYR)
168.00
EA
Add to cart
Capacity (Metric):
30 L
Dimensions (L x W x H):
400 x 335 x 318 mm
Price (MYR)
168.00
EA
Add to cart
Manufactured from Polypropylene
Includes lid
Suitable for clinical waste removal
Conforms to BS7320
Specifications
Capacity (Metric)
30 L
Certifications/Compliance
BS 7320
Color
Yellow
Dimensions (L x W x H)
400 x 335 x 318 mm
Height (Metric)
318 mm
Length (Metric)
400 mm
Width (Metric)
335 mm
Material
Polypropylene
Product Type
Waste Removal Bin