Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS15499103100 mL
FKS15409113250 mL
FKS15429113500 mL
Catalog number FKS15499103
Price (MYR)
238.00
PK
Add to cart
Capacity (Metric):
100 mL
Price (MYR)
238.00
PK
Add to cart
  • Borosilicate glass
  • Graduated
Specifications
Diameter (Metric) Outer Neck34 mm
Capacity (Metric)100 mL
MaterialBorosilicate Glass
Height (Metric)105 mm
GraduatedYes
Diameter (Metric) Outer64 mm
Unit SizePK