Squat Form Beakers
Actual product may vary
Fisherbrand
Squat Form Beakers
Constructed of borosilicate glass. Fisherbrand™ Squat Form Beakers are graduated and come in a large variety of capacities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS15409083250 mL
FKS1546907325 mL
FKS1547907350 mL
FKS15489073100 mL
FKS15499073150 mL
FKS15419083400 mL
FKS15429083600 mL
FKS154490831000 mL
FKS154590832000 mL
FKS154790835000 mL
Catalog number FKS15409083
Price (MYR)
168.00
Pack
Capacity (Metric):
250 mL
Price (MYR)
168.00
Pack
Constructed of borosilicate glass. Fisherbrand™ Squat Form Beakers are graduated and come in a large variety of capacities.
  • Borosilicate Glass 3.3
  • Low Form Graduated with Spout
  • Manufactured to ISO 3819
Specifications
ColorTranslucent
Diameter (Metric) Outer70 mm
Capacity (Metric)250 mL
MaterialBorosilicate Glass (3.3)
FinishMatte
Height (Metric)95 mm
IncludesSpout
Quantity10 Pk.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceISO 3819
ScaleWhite Graduated Scale
StyleLow Form
Unit SizePack