Samco™ Extra Long Transfer Pipettes
Samco™ Extra Long Transfer Pipettes
Actual product may vary
Fisherbrand
Samco™ Extra Long Transfer Pipettes
Perform safe and easy sampling from large containers using these extra-long transfer pipettes, perfect for research applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityPackaging TypeQuantity
FKS1539785310.7 mLSterileIndividually Wrapped100/Pk 8Pk/Cs 800/Cs
FKS153578535.6 mLNon-sterileBulk400/Pk 8Pk/Cs 4000/Cs
FIS13-711-4010.7 mLNon-sterileBulk100/Pk 10Pk/Cs 1000/Cs
263-1S10.7 mLSterileIndividually Wrapped100/Pk 8Pk/Cs 800/Cs
FIS02-708-03010.7 mLSterileIndividually Wrapped100/Pk 8Pk/Cs 800/Cs
FKS1165183923 mLNon-sterileWhite Box100/Pk.
Catalog number FKS15397853
Price (MYR)
574.00
PK
Add to cart
Volume (Metric):
10.7 mL
Sterility:
Sterile
Packaging Type:
Individually Wrapped
Quantity:
100/Pk 8Pk/Cs 800/Cs
Price (MYR)
574.00
PK
Add to cart
Perfect for research applications, these Extra-Long Transfer Pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, and autoclaving and prevents cross-contamination. They allow safe and easy sampling from large containers.
  • Excellent for research applications
  • General-purpose pipettes are ideal for routine sampling of liquids
  • Well suited for use with large containers
Specifications
Bulb Draw (Metric)10.7 mL
Diameter (Metric) Outer8.6 mm
MaterialLDPE
SterilitySterile
Drops/mL20
GraduatedNo
Length (English)12 in.
Quantity100/Pk 8Pk/Cs 800/Cs
Packaging TypeIndividually Wrapped
Volume (Metric)10.7 mL
Length (Metric)30.0 cm
TypeExtra Long Transfer Pipet