Snap-on or Push-in Cap Glass Specimen Vials
Snap-on or Push-in Cap Glass Specimen Vials
Actual product may vary
Snap-on or Push-in Cap Glass Specimen Vials
Store and display specimens securely and easily. Fisherbrand™ Snap-on or Push-in Cap Glass Specimen Vials are available in a variety of sizes. Fisherbrand™ snap-on or screw-in polyethylene caps are sold separately.
Have Questions?
Catalog NumberVolume (Metric)
FKS1539476921 mL
Catalog number FKS15394769
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
21 mL
  • Manufactured from clear Type 1B neutral glass
  • Versatile capacity range from a variety of sizes
  • Can be used with either snap-on or push-in caps
  • Rolled rim for easy cap application
  • Particularly suitable for the storage of powders and dry samples
  • Fisherbrand™ Polyethylene Push-in or Snap-on Caps for Glass Specimen Vials sold separately
Specifications
ColorClear
Diameter (Metric) Outer23 mm
MaterialNeutral Glass (Type 1B)
Volume (Metric)21 mL
Closure TypeSnap On or Push In Cap
Height (Metric)66 mm
Closure Size22 mm
Unit SizePK