Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Actual product may vary
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Samco™ Narrow Stem Transfer Pipettes
Ideal for work with narrow tubes and containers with small openings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityBulb Draw (Metric)Packaging Type
FKS153878434.5 mLNon-sterile3.3 mLBulk
FKS153178431.3 mLNon-sterile0.8 mLBulk
6946.9 mLNon-sterile3.2 mLBulk
Catalog number FKS15387843
Price (MYR)
163.00
PK
Add to cart
Volume (Metric):
4.5 mL
Sterility:
Non-sterile
Bulb Draw (Metric):
3.3 mL
Packaging Type:
Bulk
Price (MYR)
163.00
PK
Add to cart
Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Narrow Stem Transfer Pipettes. These polyethylene pipettes prevent cross-contamination and are used narrow tubes and containers with small openings . The single-unit design features a built-in pipette bulb, and a flexible stem bends to sample from narrow or small tubes.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications
Bulb Draw (Metric)3.3 mL
DescriptionSedi-Pet Transfer Pipette, Narrow Stem, Standard Bulb 15.3mL
Diameter (Metric) Outer2.5 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL26
GraduatedNo
Quantity500/Pk 10Pk/Cs 5000/Cs
Packaging TypeBulk
Volume (Metric)4.5 mL
Length (Metric)15.5 cm
TypeNarrow Stem Transfer Pipet