Delrin™ Full-Size Test Tube Racks, 24 x 30mm
Delrin™ Full-Size Test Tube Racks, 24 x 30mm
Actual product may vary
Delrin™ Full-Size Test Tube Racks, 24 x 30mm
Molded in a single, continuous piece. Fisherbrand™ Delrin™ Full-Size Test Tube Racks, 24 x 30mm are highly resistant to heat and chemical attack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS15360400Yellow
FKS10257963Blue
Catalog number FKS15360400
Price (MYR)
203.00
EA
Add to cart
Color:
Yellow
Price (MYR)
203.00
EA
Add to cart
  • Delrin™ fiberglass-reinforced polyoxymethylene (POM)
  • Offers the highest chemical and heat resistance (-40 to +140°C)
  • Unique tear drop shaped openings for better retention of tubes
  • Autoclavable
  • Molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
Specifications
Product TypeTest Tube Rack
AutoclavableAutoclavable
ColorYellow
HoldsTubes (15 to 50 mL)
MaterialFiberglass-reinforced POM
Positions24
For Use With (Equipment)15 to 50mL test tubes
No. per Case1
QuantityEach
Dimensions (L x W x H)110 x 280 x 85 mm
Length (Metric)110 mm
Width (Metric)280 mm
Height (Metric)85 mm
Diameter (Metric) Opening30 mm
Unit SizeEA