Check Order Status
Natural Polyethylene Snap-on Caps
Actual product may vary
Natural Polyethylene Snap-on Caps
Snaps onto glass rolled-rim specimen vials. Fisherbrand™ Natural Polyethylene Snap-on Caps are made of natural polyethylene.
Have Questions?
Catalog Number
Closure Size
FKS15344789
22 mm
Catalog number
FKS15344789
Price (MYR)
163.00
PK
Add to cart
Closure Size:
22 mm
Price (MYR)
163.00
PK
Add to cart
Natural polyethylene construction
Snaps onto rolled-rim specimen vials
Supplied in quantities of 300
Vials sold separately
Specifications
Closure Size
22 mm
Material
Polyethylene
Color
Natural
Format
Snap-on
Quantity
300/Pk.
For Use With (Equipment)
Glass Rolled-rim Specimen Vials