Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Actual product may vary
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Samco™ Transfer Pipettes
Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityBulb Draw (Metric)Packaging Type
FKS153078137.7 mLNon-sterile3.5 mLBulk 500/Pk
202-20S7.7 mLSterile3.5 mL20/Bag, 25 Bags/Pk, 8 Pks/Cs.
Catalog number FKS15307813
Price (MYR)
146.00
PK
Add to cart
Volume (Metric):
7.7 mL
Sterility:
Non-sterile
Bulb Draw (Metric):
3.5 mL
Packaging Type:
Bulk 500/Pk
Price (MYR)
146.00
PK
Add to cart
Prevent cross-contamination with Thermo Scientific™ Samco™ Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
Recommended for: Research Applications
Specifications
Bulb Draw (Metric)3.5 mL
DescriptionGeneral Purpose Transfer Pipettes, Standard Bulb, 7.7mL
Diameter (Metric) Outer7.9 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL21
GraduatedNo
Packaging TypeBulk 500/Pk
Volume (Metric)7.7 mL
Length (Metric)15.6 cm
TypeGeneral Purpose Transfer Pipet