Glass Screw Neck Specimen Vials
Glass Screw Neck Specimen Vials
Actual product may vary
Glass Screw Neck Specimen Vials
Store specimens securely with squat or tall form glass vials. Fisherbrand™ Glass Screw Neck Specimen Vials are available in a variety of sizes. Fisherbrand™ polypropylene screw caps are sold separately.
Have Questions?
Catalog NumberVolume (Metric)Closure SizeStyle
FKS1530478910.5 mL15 mmTall form
Catalog number FKS15304789
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
10.5 mL
Closure Size:
15 mm
Style:
Tall form
  • Manufactured from clear Type 1B neutral glass
  • Squat and tall form options with standard neck sizes
  • 1 dram equates to approximately 3.55mL (or 0.125 fl. oz.)
  • Suitable for media, diagnostic, storage, display, and sample collection applications
  • Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials sold separately
Specifications
ColorClear
Diameter (Metric) Outer17 mm
MaterialNeutral Glass (Type 1B)
Volume (Metric)10.5 mL
StyleTall form
Closure TypeScrew Cap
Height (Metric)67 mm
Closure Size15 mm
Unit SizePK