Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Actual product may vary
Fisherbrand
Bulb Pipettes, One Mark, Class AS, Certified, ISO 648, DIN 12691
Pipette with the highest precision. Fisherbrand™ one mark Class AS pipettes are made of soda lime glass. Colour coded with blue markings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FKS1526793525 mL
FKS152279351 mL
Catalog number FKS15267935
Price (MYR)
96.00
PK
Add to cart
Volume (Metric):
25 mL
Price (MYR)
96.00
PK
Add to cart
Pipette with the highest precision. Fisherbrand one mark Class AS pipettes are made of soda lime glass. Colour coded with blue markings.
  • Mark one pipet for precision measurment
  • Type one pipet with zero mark at the top
  • Soda lime glass 
  • Batch Certificate of Compliance is downloadable 
Specifications
MaterialSoda Lime Glass
GraduatedYes
Quantity2 Pack
Volume (Metric)25 mL
TypePipet
Unit SizePK