Reagent Bottle, Wide Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck
Reagent Bottle, Wide Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck
Actual product may vary
Fisherbrand
Reagent Bottle, Wide Neck, Soda-Lime Glass, Amber, Standard Taper Ground Joint Neck
Classic wide-neck, glass-stoppered reagent bottles in amber soda lime glass.
Have Questions?
Catalog NumberCapacity (Metric)
FKS15231839500 mL
Catalog number FKS15231839
Price (MYR)
4,180.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
4,180.00
PK
Add to cart
A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
  • UV protection due to external colouration
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • All glass components
  • Tight closure due to standard ground joint
Specifications
Capacity (Metric)500 mL
For Use With (Application)Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents and chemicals; Storage of hygroscopic liquids, powders and granules
IncludesWith standard ground glass flat-headed stopper
MaterialSoda Lime Glass
Quantity10/Pk.
AutoclavableNot Autoclavable
Diameter (Metric)86 mm
Unit SizePK