Check Order Status
Nichrome Inoculating Needles with Handles
Actual product may vary
Fisherbrand
Nichrome Inoculating Needles with Handles
Available in two needle gauges
Have Questions?
Change view
Catalog Number
Gauge
FKS15228979
26 G
FIS14-956-100
26 G
FIS14-956-101
24 G
3 Options
Catalog number
FKS15228979
Price (MYR)
78.00
EA
Add to cart
Gauge:
26 G
Price (MYR)
78.00
EA
Add to cart
Consist of a straight wire fixed into an aluminum handle
Handle is 7.25 in. long (18.4cm) and has a tapered end
Wire is 100mm long
Specifications
Length (English)
3.93 in.
Length (Metric)
10 cm
Material
Nichrome
Product Type
Inoculating Needle
Gauge
26 G
Length (English) Handle
7.25 in.
Length (Metric) Handle
18.4 cm
Handle Type
Aluminum handle