BMP21-LAB Portable Lab Printer Kit UK
BMP21-LAB Portable Lab Printer Kit UK
Actual product may vary
BMP21-LAB Portable Lab Printer Kit UK
BMP21-LAB Portable Lab Printer Kit UK
BMP21-LAB Portable Lab Printer Kit UK
Fisherbrand
BMP21-LAB Portable Lab Printer Kit UK
The BMP21-LAB Portable Label Printer is an all-in-one labeller for research, academic and clinical labs. Quickly and easily create legible labels for flat, curved or highly textured surfaces that stay stuck for years even when exposed to extreme temperatures and chemicals.
Have Questions?
Catalog number FKS15218957
Price (MYR)
0.00
EA
Add to cart
Price (MYR)
0.00
EA
Add to cart
  • Designed for Laboratory: additional laboratory symbols in font 9 and up, improved barcode quality, ability to preset custom label lengths
  • Labeling: materials available for applications such as cryo vials and slides; Greek symbols commonly used in laboratory, intuitive menu structure for symbols
  • Vial label function automatically cuts the tape according to the size of the vial; Time & Date stamp; Automated serialisation
  • Laboratory specific materials designed to withstand harsh laboratory environments. High quality, long lasting labels that stick and stay stuck
  • Prints durable true-sized 6 – 19 mm wide identification to maximise amount of data. Automatic label formatting for commonly used label types in laboratory
  • Long life rechargeable Lithium-ion battery - accessories sold separately. Battery included in the Starter Kit only
  • 3 line LCD display provides clear visibility
  • 2 year Warranty
Specifications
Product TypeLabel Printer Kit UK
Display TypeMonocolor LCD
Print TechnologyThermal Transfer
Width (Metric)125 mm
Height (Metric)235 mm
Depth (Metric)50 mm
Dimensions (L x W)235 x 125 mm
Dimensions (L x W x H)90 x 155 x 255 mm
Color CapabilityMonochrome
IncludesPrinter, Li-ion Battery, Power Cord, Lanyard, Quick Start Guide, M21-750-7425 label cartridge
LanguagesCroatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Slovak, Slovenian, Spanish, Swedish, Turkish
MemoryStores 12 labels
Print Speed10 mm/sec.
Resolution203 dpi
Temperature Range (Metric)5°C to 40°C
For Use With (Application)Laboratory identification
Power Supply Details6 AA Batteries, Rechargeable Battery, AC Adaptor
Unit SizeEA