BioClean™ Clearview™ Autoclavable Goggles
BioClean™ Clearview™ Autoclavable Goggles
Actual product may vary
BioClean™ Clearview™ Autoclavable Goggles
BioClean™ Clearview™ Autoclavable Goggles
BioClean™ Clearview™ Autoclavable Goggles
Constructed in super-soft thermoplastic rubber to provide unrivalled comfort, BioClean Clearview™ Autoclavable Cleanroom Goggles feature upper vents, indirect lower vents, and have a toughened anti-scratch polycarbonate lens. Provided non-sterile. To be autoclaved at 121°C/250°F for 30 minutes
Have Questions?
Catalog number FKS15208108
Price (MYR)
-
  • Non-Sterile and Sealed in a PE Bag to maintain cleanliness
  • Upper & Indirect Ventilation holes
  • EN 166:2001 Optical Class 1
  • Toughened Anti-Scratch Polycarbonate lens
  • Tested to over 40 Autoclave Cycles
  • Super-soft thermoplastic provides unrivaled comfort
  • Processed and packed in our NEBB Certified ISO Class 4 (Class 10) Cleanroom
  • Replacement lens available (BCAG-SL 19-668-191)
Specifications
Lens CoatingScratch-resistant
Certifications/ComplianceISO 9001:2015, (EU)2016/425, EN 166:2001
Product TypeAutoclavable Goggles
No. per Case12/Cs.
No. per Pack1 Ea.
Ventilation DetailsIndirect base ventilation
MaterialPolycarbonate
Unit SizePK