Check Order Status
Syringe Filter with Hose Connector Adapter
Actual product may vary
Fisherbrand
Syringe Filter with Hose Connector Adapter
For use with Twin hose connector
Have Questions?
Catalog number
FKS15193937
Price (MYR)
948.00
PK
Add to cart
Price (MYR)
948.00
PK
Add to cart
Optional venting connector with a 0.2μm syringe filter
Specifications
Includes
Hose connector adapter
Product Type
Syringe Filter