Tip for PowderThief Sampler 32mm Diameter
Tip for PowderThief Sampler 32mm Diameter
Actual product may vary
Tip for PowderThief Sampler 32mm Diameter
Tip for PowderThief Sampler 32mm Diameter
Tip for PowderThief Sampler 32mm Diameter
Tip for PowderThief Sampler 32mm Diameter
Tip for PowderThief Sampler 32mm Diameter
Extensively used throughout the pharmaceutical, chemical and food industries around the world. Sampling Systems™ Tip for PowderThief Sampler 32mm Diameter allow a single sampler to be used to accurately sample a range of different volumes.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1518686250 mL
Catalog number FKS15186862
Price (MYR)
1,144.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
1,144.00
EA
Add to cart
  • Made from high quality 316L stainless steel
  • Wide range of sample sizes
  • Wide range of sampler lengths
  • Easy to use
  • A range of tips that let a single sampler be used to accurately sample a range of different volumes
Specifications
Product TypeTip
MaterialStainless Steel (316L)
Capacity (Metric)50 mL
For Use With (Application)Powder sampling
Diameter (Metric)32 mm
Unit SizeEA