GL45 Screw cap for Pyrex GL 45 media-lab bottle
GL45 Screw cap for Pyrex GL 45 media-lab bottle
Actual product may vary
Fisherbrand
GL45 Screw cap for Pyrex GL 45 media-lab bottle
Facilitate the transfer of liquids within a closed and sterile connection system. The GL45 screw cap for Pyrex™ GL45 media-lab bottle is part of a Pyrex™ connection system perfect for chemistry or biotechnology laboratories.
Have Questions?
Catalog NumberIncludes
FKS15173927Two GL14 ports
Catalog number FKS15173927
Price (MYR)
783.00
PK
Add to cart
Includes:
Two GL14 ports
Price (MYR)
783.00
PK
Add to cart
  • Connection system screwcaps for Pyrex™ GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system, ideal for chemistry and biotechnology laboratories
  • Begin by choosing a two or three port connection system screwcap, add tubing adapters and inserts to attach tubing
  • Four different tubing diameters can be accommodated (OD 1.6mm, 3.0mm, 3.2mm and 6.0mm)
  • The additional pressure compensation set that includes a membrane filter with 0.2μ m sized pores permits sterile pressure equalisation
  • Unused ports can be sealed with a blanking cap (4538/04)
  • Components are fully interchangeable and compatible with both two and three port screwcaps
  • Screw caps and components are temperature resistance to 140°C
  • Autoclavable and dishwasher safe
  • Manufactured from polypropylene with tubing connectors that allow GL 45 media-lab bottles to be connected with flexible tubing
  • Suitable for use with soft elastic tubing that has an inner diameter of 6mm to 9mm (such as Viton™, Tygon™ or other silicon tubing)
  • Bottle contents can be kept sterile with an optional venting connector with a 0.2μ m syringe filter
  • The top angled connectors prevent kinks in the tubing
  • Screw cap can be removed without the need to remove the tubing as the centre rotates freely
  • Temperature resistance to 140°C
  • Autoclavable
  • Dishwasher safe
Specifications
AutoclavableAutoclavable
MaterialPolypropylene
For Use With (Application)Ideal for chemistry and biotechnology laboratories
Thread Size45 mm
IncludesTwo GL14 ports
Product TypeGL45 Screw Cap
Unit SizePK