Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes
Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes
Actual product may vary
Fisherbrand
Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes
Manufactured from Pyrex™ borosilicate glass and complies to DIN 12336 standards. Pyrex™ Borosilicate Glass Flat Bottom Evaporating Dishes are highly resistant to chemical erosion and thermal shock. Flat base with curved sides and spout provide easy handling and helps to prevent spillage.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS15137193320 mL115 mm65 mm
Catalog number FKS15137193
Price (MYR)
183.00
EA
Add to cart
Capacity (Metric):
320 mL
Diameter (Metric):
115 mm
Height (Metric):
65 mm
Price (MYR)
183.00
EA
Add to cart
  • Complies to DIN 12336
  • Manufactured from Pyrex™ borosilicate glass
  • High resistance to chemical erosion and thermal shock
  • Flat base
  • Curved sides with spout
Specifications
Bottom ShapeFlat
Capacity (Metric)320 mL
MaterialBorosilicate Glass
ShapeRound
TypeEvaporating Dish
Height (Metric)65 mm
Diameter (Metric)115 mm
Unit SizeEA