GL14 screw cap for Pyrex™ GL 45 media-lab bottle
GL14 screw cap for Pyrex™ GL 45 media-lab bottle
Actual product may vary
Fisherbrand
GL14 screw cap for Pyrex™ GL 45 media-lab bottle
Facilitate the transfer of liquids within a closed and sterile connection system. The GL14 screw cap for Pyrex™ GL45 media-lab bottle is part of a Pyrex™ connection system perfect for chemistry or biotechnology laboratories.
Have Questions?
Catalog NumberIncludes
FKS15123937Red PTFE coated insert
Catalog number FKS15123937
Price (MYR)
129.00
PK
Add to cart
Includes:
Red PTFE coated insert
Price (MYR)
129.00
PK
Add to cart
  • Connection system screwcaps for Pyrex™ GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system, ideal for chemistry and biotechnology laboratories
  • Begin by choosing a two or three port connection system screwcap, add tubing adapters and inserts to attach tubing
  • Four different tubing diameters can be accommodated (OD 1.6mm, 3.0mm, 3.2mm and 6.0mm)
  • The additional pressure compensation set that includes a membrane filter with 0.2μ m sized pores permits sterile pressure equalisation
  • Unused ports can be sealed with a blanking cap (15123937)
  • Components are fully interchangeable and compatible with both two and three port screwcaps
  • Screw caps and components are temperature resistance to 140°C
  • Autoclavable and dishwasher safe
Specifications
AutoclavableAutoclavable
MaterialPolypropylene
For Use With (Application)Ideal for chemistry and biotechnology laboratories
Thread Size14 mm
IncludesRed PTFE coated insert
Product TypeGL14 Screw Cap
Unit SizePK