Check Order Status
Type III Soda Lime Glass Specimen Vial
Actual product may vary
Fisherbrand
Type III Soda Lime Glass Specimen Vial
Use these dependable vials for small sample collection, analysis or storage. Fisherbrand™ Type III Soda Lime Glass Specimen Vials are ideal for a range of diagnostic, pharmaceutical and healthcare applications.
Have Questions?
Catalog Number
Volume (Metric)
FKS14851582
16 mL
Catalog number
FKS14851582
Price (MYR)
393.00
EA
Add to cart
Volume (Metric):
16 mL
Price (MYR)
393.00
EA
Add to cart
Clear Type III Soda Lime Glass vial
With push-in cap
Specifications
Diameter (Metric) Inner
25 mm
Color
Clear
Material
Soda Lime Glass (Type 3)
Volume (Metric)
16 mL
No. per Pack
100
Closure Material
Polyethylene
Closure Type
Push-in Cap
Height (Metric)
50 mm
Quantity
100 Pk.