Clear Moulded Glass Vials, with Fitted Screw Caps
Actual product may vary
Clear Moulded Glass Vials, with Fitted Screw Caps
Clear Moulded Glass Vials, with Fitted Screw Caps
Clear Moulded Glass Vials, with Fitted Screw Caps
Suitable for the storage of pathology media. Fisherbrand™ Clear Moulded Glass Vials, with Fitted Screw Caps are manufactured from clear Type 1 neutral glass.
Have Questions?
Catalog NumberVolume (Metric)Closure Material
FKS1480357228 mLAluminum
Catalog number FKS14803572
Price (MYR)
344.00
Pack
Volume (Metric):
28 mL
Closure Material:
Aluminum
Price (MYR)
344.00
Pack
  • With screw neck
  • Autoclavable
  • Available in a range of capacities
  • With either fitted aluminum rubber lined cap or PP cap
Specifications
Material (Liner)Rubber
ColorClear
Diameter (Metric) Outer28 mm
MaterialBorosilicate Glass
Volume (Metric)28 mL
Neck StyleScrew
Thread StyleScrew
Closure MaterialAluminum
Closure TypeScrew Cap
Quantity20/Pk.
Closure Size28 mm
AutoclavableAutoclavable
Height (Metric)83 mm
Product TypeUniversal Vial
Unit SizePack