Sterile Graduated Transfer Pipets
Sterile Graduated Transfer Pipets
Actual product may vary
Fisherbrand
Sterile Graduated Transfer Pipets
Made up of low density polyethylene.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)Volume (Metric) DispensePackaging Type
FKS134791083 mL155 mm40 μL50 inner packs of 10
FKS134691083 mL155 mm40 μLIndividually Wrapped
FKS134091183 mL155 mm40 μL25 inner packs of 20
Catalog number FKS13479108
Price (MYR)
208.00
PK
Add to cart
Volume (Metric):
3 mL
Length (Metric):
155 mm
Volume (Metric) Dispense:
40 μL
Packaging Type:
50 inner packs of 10
Price (MYR)
208.00
PK
Add to cart
  • Transparent in color
  • Contains graduations
  • Sterile
Specifications
MaterialLDPE
Drops/mL25
GraduatedYes
Graduations0.5 mL
Quantity500/Pk.
Volume (Metric) Dispense40 μL
Packaging Type50 inner packs of 10
Volume (Metric)3 mL
Length (Metric)155 mm
TypeTransfer Pipet
Unit SizePK