Non-sterile Transfer Pipettes, Transparent LDPE
Non-sterile Transfer Pipettes, Transparent LDPE
Actual product may vary
Fisherbrand
Non-sterile Transfer Pipettes, Transparent LDPE
Prevent cross contamination. Fisherbrand™ Non-sterile Transfer Pipets, Transparent LDPE are made of shatterproof, flexible plastic and are designed to replace glass Pasteur pipets. Graduated and non-graduated pipets available in a range of sizes and styles, including jumbo, thin stem and extra long.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Drops/mLGraduatedTip Style
FKS134491087 mL20NoExtra Long
FKS134391083 mL25YesStandard
Catalog number FKS13449108
Price (MYR)
49.00
PK
Add to cart
Volume (Metric):
7 mL
Drops/mL:
20
Graduated:
No
Tip Style:
Extra Long
Price (MYR)
49.00
PK
Add to cart
  • Built-in bulb eliminates the need for plugging, packaging and autoclaving
  • Flexible stem bends to sample from narrow or small tubes
  • Low affinity surface reduces loss of cells and valuable proteins due to binding
  • Soft plastic pipets made of non-toxic low-density polyethylene (LDPE)
  • Non-sterile, transparent and fully recyclable
Specifications
MaterialLDPE
Drops/mL20
GraduatedNo
Quantity100 Pack
Tip StyleExtra Long
Volume (Metric) Dispense50 μL drop
Volume (Metric)7 mL
Length (Metric)300 mm
TypeTransfer Pipet
Unit SizePK