Wide-Form Porcelain Crucibles
Wide-Form Porcelain Crucibles
Actual product may vary
Fisherbrand
Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) OuterHeight (Metric)Weight (Metric)
FKS1342971930 mL50mm31mm26g
FISFB965K30 mL50mm31mm26g
FISFB965L50 mL61mm37mm35g
FISFB965P250 mL102mm60mm124g
FISFB965N150 mL84mm52mm90g
Catalog number FKS13429719
Price (MYR)
373.00
PK
Add to cart
Capacity (Metric):
30 mL
Diameter (Metric) Outer:
50mm
Height (Metric):
31mm
Weight (Metric):
26g
Price (MYR)
373.00
PK
Add to cart
Withstand temperatures up to 1150°C (2102°F)
  • Glazed inside and out except for outside bottom surface and rim
Specifications
MaterialPorcelain
FinishGlazed
Max. Temperature (Metric)1150°C
Thickness (Metric)2mm
Weight (Metric)26g
Capacity (Metric)30 mL
Diameter (Metric) Outer50mm
Height (Metric)31mm
Product TypeCrucible
Unit SizePK