PPCO Rectangular Carboy with Spigot
PPCO Rectangular Carboy with Spigot
Actual product may vary
PPCO Rectangular Carboy with Spigot
PPCO Rectangular Carboy with Spigot
PPCO Rectangular Carboy with Spigot
PPCO Rectangular Carboy with Spigot
PPCO Rectangular Carboy with Spigot
Fisherbrand
PPCO Rectangular Carboy with Spigot
For Use With: Ideal for storing and dispensing lab water and reagents; Autoclavable; Closure Material: PPCO; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS134180399 L
FIS03-007-64820 L
FKS1342803920 L
Catalog number FKS13418039
Price (MYR)
774.00
PK
Add to cart
Capacity (Metric):
9 L
Price (MYR)
774.00
PK
Add to cart
Value your lab can rely on. Fisherbrand PPCO Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Ideal for storing and dispensing lab water and reagents
  • Durable construction for long service life
  • Molded graduations for 1 gallon and 51 increments
  • Choice of materials for general use or autoclavability
  • Space-saving rectangular design
  • Wide mouth closure allows for easy filling and cleaning access
  • Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
  • Spigot spout without the adapter fits 5/8-in. ID tubing
  • Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
Specifications
AutoclavableAutoclavable
Capacity (English)2.4 gal.
Capacity (Metric)9 L
Certifications/ComplianceFDA, USP VI
Closure MaterialPPCO
ColorWhite
Dimensions (L x W x H)External MaxD (with spigot) x W x H (with closure): 10.5 in. x 6.0 x 14.2 in. (26.7 x 15.3 x 36.1 cm)
For Use With (Application)Ideal for storing and dispensing lab water and reagents
MaterialPPCO
Mouth StyleWide
Product TypeCarboy with Spigot
ShapeRectangular
Unit SizePK