ZX3 Vortex Mixer
ZX3 Vortex Mixer
Actual product may vary
ZX3 Vortex Mixer
ZX3 Vortex Mixer
Fisherbrand
ZX3 Vortex Mixer
Offers excellent flexibility and versatility and provides stable mixing at high speeds. Fisherbrand™ ZX3 Vortex Mixer can be connected to many platforms and accessories which are easily attached and securely fitted with simple pressure.
Have Questions?
Catalog number FKS13214789
Price (MYR)
2,778.00
EA
Add to cart
Price (MYR)
2,778.00
EA
Add to cart
Universal Operation:
  • Variable speed operation, electronically controlled up to 3000rpm
  • Automatic start in touch mode or continuous operation
  • Large choice of accessories to suit tubes, plates and flasks
  • Universal voltage / frequency model
Reliable Performance:
  • Accessories fit by simple pressure
  • Optimum resistance to chemicals from technopolymer casing
  • Excellent stability with zinc alloy base
  • Large, anti-slip rubber feet for staying in required position
  • IP 42 protection against particles and liquids
Specifications
Orbit4.2 mm
Speed Range0 to 3000 rpm
Electrical Requirements100/240 V, 50/60 Hz, 15 W
Voltage100/240 V
Wattage15 W
Product TypeVortex Mixer
Frequency50/60 Hz
MaterialZinc Alloy, Technopolymer
Depth (Metric)165 mm
Height (Metric)130 mm
Width (Metric)150 mm
Weight (Metric)2.7 kg
ModelZX3
IP RatingIP 42
Operating ModesTouch, Continuous
Unit SizeEA