Unisex Cotton Laboratory Coats
Unisex Cotton Laboratory Coats
Actual product may vary
Fisherbrand
Unisex Cotton Laboratory Coats
Combine professionalism and function with Logistik™ Unisex Cotton Laboratory Coats. Available in sizes small through 2XL, the coats are all cotton, unisex.and feature a back vent for a comfortable fit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeSize (Metric) Chest
FKS13181731X-Large116.84 to 121.92 cm
FKS13161731Medium101.6 to 104.14 cm
FKS13151731Large106.68 to 111.76 cm
FKS13171731Small91.44 to 96.52 cm
Catalog number FKS13181731
Price (MYR)
506.00
EA
Add to cart
Size:
X-Large
Size (Metric) Chest:
116.84 to 121.92 cm
Price (MYR)
506.00
EA
Add to cart
  • Unisex
  • Pure cotton
  • Concealed stud fastened front
  • 305g2 heritage 3111 cotton
  • Not CE marked
Specifications
Product TypeGeneral Purpose Lab Coat
SizeX-Large
GenderUnisex
MaterialCarbon
ColorWhite
DisposableReusable
SterilityNon-sterile
No. of Pockets3
Size (Metric) Chest116.84 to 121.92 cm
IncludesThree mitred patch pockets
Length (Metric)105 cm
Construction100% Cotton
Unit SizeEA