Soda Lime Glass Watch Glasses
Soda Lime Glass Watch Glasses
Actual product may vary
Fisher Catalogue
Soda Lime Glass Watch Glasses
Manufactured from Soda lime glass. Scientific Glass Labs™ Soda Lime Glass Watch Glasses with thin, clear glass and ground edge can be used to evaporate liquids, hold solids for weighing, or for heating small substances.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1317243040 mm
FKS13122440100 mm
FKS13142440150 mm
FKS1310244080 mm
FKS1311244090 mm
Catalog number FKS13172430
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
40 mm
Watch glasses are useful for evaporating liquids, holding solids for weighing, or for heating small substances.
  • Manufactured from Soda lime glass
  • Thin, clear glass
  • Ground edge
Specifications
Bottom ShapeFlat
MaterialSoda Lime Glass
ShapeRound
TypeWatch Glass
For Use With (Application)Evaporating Liquids, Heating Small Substances, Holding Solids for Weighing
Interior FinishGround Edge
Diameter (Metric)40 mm
Unit SizePK